Learn More
Qosina Corp CHECK VALVE, F INLET, M OUTLET
Catalog No. NC1769618
Encompass
Qosina offers an extensive line of Check Valves, available in a variety of styles that are in stock and ready for immediate delivery. Qosina part #80107 features a female luer lock inlet and male luer lock outlet. Made of polycarbonate and silicone, this in-line valve is tinted for gamma sterilization and has a 1.5 - 5 psi cracking pressure range, determining when the component starts to open. Qosina offers a variety of flow rates, materials, cracking pressure and back pressure options. Check Valve sample kits are also available which are ideal for testing.
Catalog No. NC1769618 Supplier Qosina Corp Supplier No. 80107
Qosina offers an extensive line of Check Valves, available in a variety of styles that are in stock and ready for immediate delivery. Qosina part #80107 features a female luer lock inlet and male luer lock outlet. Made of polycarbonate and silicone, this in-line valve is tinted for gamma sterilization and has a 1.5 - 5 psi cracking pressure range, determining when the component starts to open. Qosina offers a variety of flow rates, materials, cracking pressure and back pressure options. Check Valve sample kits are also available which are ideal for testing.