Learn More
Chemglass Life Sciences Chemglass Polycarbonate Check Valve, Female to Male Luer Lock (CLS-1399-F55)
SDP
Catalog No. 502733301
Encompass
The Chemglass Polycarbonate Check Valve (CLS-1399-F55) is a compact one-way valve designed for use with bioreactors and laboratory tubing sets. It connects a female luer lock to a male luer lock to provide reliable unidirectional flow control and help prevent backflow during fluid transfer and sampling.
  • Made from polycarbonate for chemical resistance and visual clarity.
  • Connects a female luer lock to a male luer lock.
  • Provides one-way flow control to prevent backflow.
  • Intended for bioreactor and laboratory tubing set applications.
Catalog No. 50-273-3301 Supplier Chemglass Life Sciences Supplier No. CLS1399F55
The Chemglass Polycarbonate Check Valve (CLS-1399-F55) is a compact one-way valve designed for use with bioreactors and laboratory tubing sets. It connects a female luer lock to a male luer lock to provide reliable unidirectional flow control and help prevent backflow during fluid transfer and sampling.
  • Made from polycarbonate for chemical resistance and visual clarity.
  • Connects a female luer lock to a male luer lock.
  • Provides one-way flow control to prevent backflow.
  • Intended for bioreactor and laboratory tubing set applications.