Learn More
Perkin Elmer US LLC Check Valve Kit for AA Furnace Autosampler
Catalog No. 502117310
Encompass_Preferred
The check valves are compatible with the following PerkinElmer furnace autosamplers: AS-900, AS-800, AS-72, AS-71, AS-70 and AS-60.
Catalog No. 50-211-7310 Supplier Perkin Elmer US LLC Supplier No. B3001485
The check valves are compatible with the following PerkinElmer furnace autosamplers: AS-900, AS-800, AS-72, AS-71, AS-70 and AS-60.