Learn More
Perkin Elmer US LLC Check Valve Kit for ASXPRESS PLUS
Catalog No. 502111326
Encompass_Preferred
The value body is polypropylene and the diaphragm is Viton. The two barbed ends are 3.2 mm in diameter.
Catalog No. 50-211-1326 Supplier Perkin Elmer US LLC Supplier No. N0777560
The value body is polypropylene and the diaphragm is Viton. The two barbed ends are 3.2 mm in diameter.