Learn More
Grainger Parker C Series Check Valve, Steel, 3/8 in. NPT, 5,000 psi, 8 gpm
Catalog No. 502794367
Encompass_Preferred
These C Series check valves are engineered for uni-directional flow control of liquids and gases across various industrial sectors, including chemical processing, oil and gas, and power generation. Constructed from steel with a black oxide finish, this inline valve features a 3/8 in. NPT connection and is designed to handle pressures up to 5,000 psi and flow rates up to 8 GPM.
  • Resilient, custom molded, blow-out resistant seat design
  • Back stopped poppet minimizes spring stress
  • 100% factory tested for both crack and reseat
  • Heat code traceability
  • Suitable for oil media
Catalog No. 50-279-4367 Supplier Grainger Supplier No. 1A045
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
These C Series check valves are engineered for uni-directional flow control of liquids and gases across various industrial sectors, including chemical processing, oil and gas, and power generation. Constructed from steel with a black oxide finish, this inline valve features a 3/8 in. NPT connection and is designed to handle pressures up to 5,000 psi and flow rates up to 8 GPM.
  • Resilient, custom molded, blow-out resistant seat design
  • Back stopped poppet minimizes spring stress
  • 100% factory tested for both crack and reseat
  • Heat code traceability
  • Suitable for oil media