Learn More
Chemglass Life Sciences Vacuum Check Valve
SDP
Catalog No. CG453901
Encompass_Preferred
Vacuum Check Valve. Valve has an 1/4in O.D. precision ground ceramic ball which fits into the ground seat to effect a liquid seal. Tubing O.D. is 10mm while the overall length is approx. 125mm.
Catalog No. CG453901 Supplier Chemglass Life Sciences Supplier No. CG453901
Vacuum Check Valve. Valve has an 1/4in O.D. precision ground ceramic ball which fits into the ground seat to effect a liquid seal. Tubing O.D. is 10mm while the overall length is approx. 125mm.