Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Phoenix™ Checkerspot Challenge Deluxe Kit Components
SCI_ED

Catalog No. S39370KCC Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Trash bags
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S39370KCC Trash bags
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S39370KCC
Add to Cart
Add to Cart

Replacement parts for Educational Kit

Variety of components specially formatted as replacements for AquaPhoenix custom designed hands-on educational kits.

Specifications

Color Black
Grade K to 5
Quantity 50/Roll
Capacity (English) 7 gal.
Thickness (English) 0.23 mil
Product Type Trash bags
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.