Learn More
Next Advance Inc Checkit Go 50μl, Box of 6
SDP
Supplier:  Next Advance Inc CK8C506
Encompass
The Checkit Go enables you to easily measure the volumes dispensed by a robot or multi-channel pipette, in under 10 seconds. Just place the dye in the reservoir on the cartridge, and place the well-plate-size cartridge on the robot (or where you are using the multi-channel pipette. Dispense the dye into the 8 wells, flip the tab, and read the level of the dye. It’s that easy!
Catalog No. NC2076903