Learn More
Lovibond™ CHECKIT™ Comparator Test Kits: Total Hardness HR
Catalog No. 09053134 Shop All Tintometer Group Products
Change view
Click to view available options
Range:
5 to 500mg/L
Catalog No. Range
09-053-134 5 to 500mg/L
1 options
Catalog No. 09-053-134 Supplier Lovibond™ Supplier No. 414240
Add to Cart
Add to Cart
Accurate test kit for routine water analysis
Specifications
For Use With (Application) Routine water analysis
Includes 30mL calibrated sample container, 30mL calibrated sample container, total hardness tablets, instructions, storage case
No. of Tests 100
Range 5 to 500mg/L
Type Total Hardness Mini Kit