Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ CHECKIT™ Comparator Test Kits

Catalog No. 09053155 Shop All Tintometer Group Products
Change view
Click to view available options
Range:
0 to 5mg/L
Type:
Bromine Color Disc
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range Type
09-053-155 0 to 5mg/L Bromine Color Disc
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-053-155 Supplier Lovibond™ Supplier No. 147280
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Carrying case, comparator, color disc, sample cells, instructions and enough reagents for 100 tests

Accurate test kits for routine water analysis

  • Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value
  • Color discs and reagents are available separately

Specifications

For Use With (Application) For routine water analysis
For Use With (Equipment) Aqua comparator™
Range 0 to 5mg/L
Sufficient For 100 Tests
Type Bromine Color Disc
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.