Description
Perform water analysis quickly and easily. Lovibond Checkit™ Comparitor Test Kits are highly accurate and incorporate a user-friendly colorimetric scale. Each kit indicates a combination of pH and another measure.
- Easy operation and exact reagent dosing
- Tablet reagents with a minimm guaranteed shelf life of 5/10 years
- Rotate continuous gradient color wheel to obtain a color match with the reacted sample
- Printed continuous gradient color discs provide accurate measurements without "exrapolating" between discrete color blocks
Specifications
Specifications
| Description | DEHA |
| Range | 0 to 0.5 mg/L |
| Test Method | Colorimetric |
| Type | Water test kit |