Learn More
MCR Safety™ Checklite CL1 Gray UV-AF Lens
Catalog No. 19034407 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Lens Coating:
Anti-fog, Impact-resistant
Catalog No. Lens Coating
19-034-407 Anti-fog, Impact-resistant
1 options
Catalog No. 19-034-407 Supplier MCR Safety™ Supplier No. CL112AF
Checklite™ CL1 series safety glasses with gray lens, UV-AF™ anti-fog coating, excellent orbital seal and fit.
  • Exclusive UV-AF™ anti-fog lens coating
  • Good for everyday use or a new visitor spec
  • Lightweight glasses (22.9 grams) are good for long term wear
  • Rugged polycarbonate lens filters 99.9% of harmful U.V. rays
  • Smaller frame sized to fit smaller faces
  • Meets ANSI/ISEA Z87.1 2020 standard
Specifications
Lens Coating Anti-fog, Impact-resistant
Lens Tint Gray
Product Type Glasses
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection U6
Frame Type Frameless
Autoclavable Not Autoclavable