Learn More
Coastline Global™ CheckUp Dog and Cat Urine Testing Strips for Detection of Diabetes, 50 count
Catalog No. 01671411
Change view
Click to view available options
Detectable Analytes:
Glucose
Catalog No. Detectable Analytes
01-671-411 Glucose
1 options
Catalog No. 01-671-411 Supplier Coastline Global™ Supplier No. CUBG
Add to Cart
Add to Cart
CheckUp dog and cat urine glucose level test strips to monitor pet's diabetes.
50 CheckUp dog or cat urine glucose test strips for diabetes detection and monitoring.
Specifications
CE Marker Self-declared
Clia Complexity Waived (Urine)
Detectable Analytes Glucose
Disposable Single-use
Form Ready-to-Use (RTU)