Learn More
Coastline Global™ CheckUp Dog and Cat Urine Testing Strips for Detection of Kidney Condition, 50 count
Catalog No. 01671403
Change view
Click to view available options
Detectable Analytes:
Protein
Catalog No. Detectable Analytes
01-671-403 Protein
1 options
Catalog No. 01-671-403 Supplier Coastline Global™ Supplier No. CUBP
Add to Cart
Add to Cart
Check for protein levels and monitor your pet's kidney condition with CheckUp urine testing strips
50 CheckUp strips for detection of protein, and indication of kidney failure, in dog or cat urine.
Specifications
CE Marker Self-declared
Clia Complexity Waived (Urine)
Detectable Analytes Protein
Disposable Single-use
Form Ready-to-Use (RTU)