Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coastline Global™ CheckUp Dog and Cat Urine Testing Strips for Detection of UTI, 50 count

Catalog No. 01671405
Change view
Click to view available options
Detectable Analytes:
pH
Catalog No. Detectable Analytes
01-671-405 pH
1 options

Catalog No. 01-671-405

Supplier: Coastline Global™ CUBU

Only null left
Add to Cart
Add to Cart

CheckUp urinary tract infection test strips for dogs and cat check for leukosytes, nitrites and pH levels to confirm the presence of and monitor UTIs.

Checks for leukosytes, nitrites and pH levels to confirm the presence of and monitor UTIs in your pet's urine.

  • 50 count of test strips
  • Can be used for dogs or cats

Specifications

CE Marker Self-declared
Clia Complexity Waived (Urine)
Detectable Analytes pH
Disposable Single-use
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.