Learn More
Coastline Global™ CheckUp Kit4Cat: Litter for feline urine sample collection
Catalog No. 01671408
Change view
Click to view available options
Detectable Analytes:
None
Catalog No. Detectable Analytes
01-671-408 None
1 options
Catalog No. 01-671-408 Supplier Coastline Global™ Supplier No. K4C
Add to Cart
Add to Cart
Kit4Cat is a veterinarian-approved revolutionary hydrophobic litter that keeps cat’s urine on top, making sample collection easy and stress free.It is non-invasive, biodegradable and 100% natural. Because it looks, feels, and smells like regular sand, cats love it.
Veterinarian-approved revolutionary hydrophobic litter that keeps cat urine on top, allowing for the easy and stress-free collection.
  • Non-invasive, biodegradable and 100% natural
  • Looks, feels, and smells like regular sand
  • 3 packs of of 300g hydrophobic sand
  • 3 disposable pipettes and vials
Specifications
CE Marker Self-declared
Clia Complexity Waived (Urine)
Detectable Analytes None
Disposable Single-use
Form Ready-to-Use (RTU)