Learn More
Bioworld Chee and Pool (C2D) Vitis Medium, 25 L
Catalog No. 50254556
Encompass_Preferred
Chee and Pool (C2D) Vitis MediumContains Macronutrients and Micronutrients as described by Chee & Pool (1987)Provided as 1 unit to make 25L or 50L of medium, or as 10 units that each make 1L of medium Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumWorking Concentration: 4.49 g/LpH: 3.5−4.5Soluble in water
Catalog No. 50-254-556 Supplier Bioworld Supplier No. 306300052
Chee and Pool (C2D) Vitis MediumContains Macronutrients and Micronutrients as described by Chee & Pool (1987)Provided as 1 unit to make 25L or 50L of medium, or as 10 units that each make 1L of medium Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumWorking Concentration: 4.49 g/LpH: 3.5−4.5Soluble in water