Learn More
Bioworld Chee and Pool (C2D) Vitis Medium w/ Vitamins, 25 L
Catalog No. 50254559
Encompass_Preferred
Chee and Pool (C2D) Vitis MediumContains Macronutrients, Micronutrients, and Vitamins as described by Chee & Pool (1987)Provided as 1 unit to make 25L or 50L of medium, or as 10 units that each make 1L of medium Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested
Catalog No. 50-254-559 Supplier Bioworld Supplier No. 306300062
Chee and Pool (C2D) Vitis MediumContains Macronutrients, Micronutrients, and Vitamins as described by Chee & Pool (1987)Provided as 1 unit to make 25L or 50L of medium, or as 10 units that each make 1L of medium Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 25L or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested