Learn More
Electron Microscopy Sciences Cheese Cloth 60 YD
SDP
Catalog No. 50366357
Encompass_Preferred
Cotton woven cloth in the pattern of 44x36 (44 lines/inch horizontally and 36 lines/inch vertically) which makes it suitable for cleaning and polishing. Supplied in 60 yard rolls (continuous) 36" in width. This configuration allows you to cut out the exact length you desire.
This item is not returnable. View return policy
Catalog No. 50-366-357 Supplier Electron Microscopy Sciences Supplier No. 7174800
Cotton woven cloth in the pattern of 44x36 (44 lines/inch horizontally and 36 lines/inch vertically) which makes it suitable for cleaning and polishing. Supplied in 60 yard rolls (continuous) 36" in width. This configuration allows you to cut out the exact length you desire.