Learn More
Lions Services Inc CHEESECLOTH GRADE90 2PLY 50YDS
Catalog No. 5039511
Encompass_Preferred
100% Cotton, Bleached, Grade 90, 2-ply, 50 yards per bolt. 36 inch width folded once to make 18 inch width 2-ply bolts. 40 warp yarns by 33 fill yarns; minimum.
This item is not returnable. View return policy
Catalog No. 50-395-11 Supplier Lions Services Inc Supplier No. 8305002623321
100% Cotton, Bleached, Grade 90, 2-ply, 50 yards per bolt. 36 inch width folded once to make 18 inch width 2-ply bolts. 40 warp yarns by 33 fill yarns; minimum.