Learn More
Lions Services Inc CHEESECLOTH GRD50 2PLY 100YDS
Catalog No. 5039508
Encompass_Preferred
Grade 50, 2-ply, 100 yards Cheesecloth. 100% Cotton, Bleached, Grade 50, 2-ply, 100 yards per bolt. 36 inch width folded once to make 18 inch width 2-ply bolts. 26 warp yarns by 22 fill yarns; minimum.
This item is not returnable. View return policy
Catalog No. 50-395-08 Supplier Lions Services Inc Supplier No. 8305002053495
Grade 50, 2-ply, 100 yards Cheesecloth. 100% Cotton, Bleached, Grade 50, 2-ply, 100 yards per bolt. 36 inch width folded once to make 18 inch width 2-ply bolts. 26 warp yarns by 22 fill yarns; minimum.