Learn More
Aviva Systems Biology Corp CHEK1 Antibody - middle region (ARP32589_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32589_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHEK1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human CHEK1. Alternate names: CHK1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54667