Learn More
Aviva Systems Biology Corp CHEK2 Antibody - N-terminal region (ARP30335_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30335_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CHEK2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CHEK2. Alternate names: CDS1, CHK2, HuCds1, LFS2, PP1425, RAD53, hCds1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-54157