Learn More
Sigma Aldrich Fine Chemicals Biosciences Chelating SepharoseR fast
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17057501
Encompass
Chelating Sepharose(R) fast flow is a BioProcess IMAC medium designed for purifying native and recombinant proteins and peptides that have an affinity for metal ions.Chelating Sepharose(R) fast flow is composed of cross-linked 6% agarose beads modified with iminodiacetic immobilized to the base matrix by stable ether linkages and sufficiently long spacer arms.Chelating Sepharose(R) fast flow is supplied free of metal ions allowing the user to charge it with the most appropriate metal ion for purification of a target protein.Chelating Sepharose(R) fast flow is available in a range of different bulk pack sizes for easy scale-up and process development. As member of the BioProcess media range Chelating Sepharose(R) fast flow meets industrial demands with security of supply and comprehensive technical and regulatory support
Catalog No. 50-290-4322