Learn More
Cayman Chemical Cheldonn 10mg
SDP
Catalog No. 502935407
Encompass_Preferred
A benzophenanthridine alkaloid; induces Bcl-2- and caspase-dependent apoptosis in HepG2 human liver carcinoma cells (LD50 = 12 µM) and in Jurkat human T cells at 1 µM; downregulates hTERT expression, reduces telomerase activity, and induces cell senescence in HepG2 cells
Catalog No. 50-293-5407 Supplier Cayman Chemical Supplier No. 2509710 MG
A benzophenanthridine alkaloid; induces Bcl-2- and caspase-dependent apoptosis in HepG2 human liver carcinoma cells (LD50 = 12 µM) and in Jurkat human T cells at 1 µM; downregulates hTERT expression, reduces telomerase activity, and induces cell senescence in HepG2 cells