Learn More
Cayman Chemical Chelerythrine (chloride), Purity: ≥98%, CAS: 3895-92-9, 10 mg
SDP
Catalog No. 502951579
Encompass_Preferred
A potent, cell permeable inhibitor of PKC (IC50 = 660 nM); also inhibits Bcl-xL function (IC50 = 1.5 μM), inducing apoptosis in several cancer cell lines; can have PKC-independent effects, activate p38 and JUNK signaling pathways, and induce apoptosis in cancer cells both in vitro and in vivo
Catalog No. 50-295-1579 Supplier Cayman Chemical Supplier No. 1131410 MG
A potent, cell permeable inhibitor of PKC (IC50 = 660 nM); also inhibits Bcl-xL function (IC50 = 1.5 μM), inducing apoptosis in several cancer cell lines; can have PKC-independent effects, activate p38 and JUNK signaling pathways, and induce apoptosis in cancer cells both in vitro and in vivo