Learn More
Selleck Chemical LLC Chelerythrine Chloride (NSC 646662) S1292-25mg
SDP
Catalog No. 503111110
Encompass_Preferred
Chelerythrine Chloride (NSC 646662 Broussonpapyrine) is a potent selective antagonist of PKC with IC50 of 0 66 M
Catalog No. 50-311-1110 Supplier Selleck Chemical LLC Supplier No. S129225MG
Chelerythrine Chloride (NSC 646662 Broussonpapyrine) is a potent selective antagonist of PKC with IC50 of 0 66 M