Learn More
Enzo Life Sciences Chelerythrine chloride (25mg). CAS: 3895-92-9
SDP
Supplier:  Enzo Life Sciences BMLEI2250025
Encompass_Preferred
Reported to be a potent and selective inhibitor of protein kinase C (IC50=0.66µM). Although chelerythrine has numerous documented biological effects, studies have raised serious doubts about the effectiveness of chelerythrine as a PKC inhibitor. Purity: ≥98% (HPLC, TLC). Solubility: Soluble in DMSO, water (2mg/ml warm), 100% ethanol or methanol. Long Term Storage: -20°C.
Catalog No. 50-201-0617