Learn More
TARGETMOL CHEMICALS INC CHELERYTHRINE CHLORIDE 25MG
SDP
Catalog No. 502580276
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Chelerythrine Chloride is a cell-permeable inhibitor of protein kinase C competitive with respect to the phosphate acceptor and non-competitive with respect to ATP. purity: 99%
Catalog No. 50-258-0276 Supplier TARGETMOL CHEMICALS INC Supplier No. T341925MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Chelerythrine Chloride is a cell-permeable inhibitor of protein kinase C competitive with respect to the phosphate acceptor and non-competitive with respect to ATP. purity: 99%