Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chelerythrine chloride | 3895-92-9 | >98.0% | 50 MG
SDP

Catalog No. 5000234122
Encompass_Preferred

Chelerythrine chloride is a cell-permeable inhibitor of protein kinase C used in biochemical and cell biology research to study PKC-dependent signaling, apoptosis, and autophagy. It is supplied as a research-grade chemical with high purity.

  • Cell-permeable protein kinase C inhibitor.
  • Reported PKC IC50 660 nM.
  • Inhibits Bcl-XL-Bak BH3 peptide binding (IC50 1.5 μM).
  • Purity >98% by HPLC.
  • Molecular formula C21H18ClNO4; molecular weight ~383.8 g/mol.
  • Supplied as a 50 mg vial for research use.

Catalog No. 50-002-34122 Supplier Medchemexpress LLC Supplier No. HY1204850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chelerythrine chloride is a cell-permeable inhibitor of protein kinase C used in biochemical and cell biology research to study PKC-dependent signaling, apoptosis, and autophagy. It is supplied as a research-grade chemical with high purity.

  • Cell-permeable protein kinase C inhibitor.
  • Reported PKC IC50 660 nM.
  • Inhibits Bcl-XL-Bak BH3 peptide binding (IC50 1.5 μM).
  • Purity >98% by HPLC.
  • Molecular formula C21H18ClNO4; molecular weight ~383.8 g/mol.
  • Supplied as a 50 mg vial for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.