Learn More
Medchemexpress LLC Chelerythrine chloride | 3895-92-9 | 99.1% | 100 MG
SDP
Catalog No. 5000231852
Encompass_Preferred
Chelerythrine chloride is a cell-permeable benzophenanthridine alkaloid used as a research reagent to inhibit protein kinase C (PKC) and to probe apoptosis and Bcl-2 family interactions in cell-based and biochemical assays.
  • Potent PKC inhibitor (IC50 660 nM).
  • Inhibits Bcl-XL-Bak BH3 peptide binding (IC50 ~1.5 μM).
  • High reported purity (99.11%).
  • Supplied as a solid powder; easily soluble in DMSO (4.35 mg/mL).
  • Recommended storage: 4°C sealed; in solution store at -80°C for up to 6 months.
Catalog No. 50-002-31852 Supplier Medchemexpress LLC Supplier No. HY12048100MG
Chelerythrine chloride is a cell-permeable benzophenanthridine alkaloid used as a research reagent to inhibit protein kinase C (PKC) and to probe apoptosis and Bcl-2 family interactions in cell-based and biochemical assays.
  • Potent PKC inhibitor (IC50 660 nM).
  • Inhibits Bcl-XL-Bak BH3 peptide binding (IC50 ~1.5 μM).
  • High reported purity (99.11%).
  • Supplied as a solid powder; easily soluble in DMSO (4.35 mg/mL).
  • Recommended storage: 4°C sealed; in solution store at -80°C for up to 6 months.