Learn More
Medchemexpress LLC HY-W016349 100mg Medchemexpress, Chelidamic acid CAS:138-60-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW016349100MG
Encompass_Preferred
Medchemexpress, HY-W016349 100mg Chelidamic acid CAS:138-60-3 Chelidamic acid is a heterocyclic organic acid with a pyran skeleton. Chelidamic acid has good coordination ability with noble metal ions. Chelidamic acid is also one of the most potent inhibitors of glutamate decarboxylase, with a K i of 33 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4616
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.