Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chelidamic acid | 138-60-3 | MFCD00066478 | 99.9% | 183.12 g/mol | C7H5NO5 | 5 G
SDP

Catalog No. 5000303255
Encompass_Preferred

Chelidamic acid is a heterocyclic organic acid used as a research reagent and an amino acid decarboxylase inhibitor. It is supplied as a high-purity solid for laboratory use in biochemical and coordination chemistry studies.

  • Small heterocyclic acid with formula C7H5NO5 and molecular weight 183.12 g/mol.
  • High purity (99.9%).
  • Soluble in DMSO and water.
  • Available in multiple laboratory pack sizes, including 5 g.
  • Suitable for biochemical assays and coordination chemistry research.

Catalog No. 50-003-03255 Supplier Medchemexpress LLC Supplier No. HYW0163495G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chelidamic acid is a heterocyclic organic acid used as a research reagent and an amino acid decarboxylase inhibitor. It is supplied as a high-purity solid for laboratory use in biochemical and coordination chemistry studies.

  • Small heterocyclic acid with formula C7H5NO5 and molecular weight 183.12 g/mol.
  • High purity (99.9%).
  • Soluble in DMSO and water.
  • Available in multiple laboratory pack sizes, including 5 g.
  • Suitable for biochemical assays and coordination chemistry research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.