Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Chelidamic acid monohydrate, 98%

Catalog No. AC108470050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108470050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC108470050 Supplier Thermo Scientific Chemicals Supplier No. 108470050
  • $197.72
    $168.65 / Each of 1

    Save $29.07

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 199926-39-1 | C7H5NO5·H2O | 201.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 199926-39-1
Molecular Formula C7H5NO5·H2O
Molecular Weight (g/mol) 201.14
MDL Number MFCD03818117
InChI Key SNGPHFVJWBKEDG-UHFFFAOYSA-N
Synonym 4-oxo-1,4-dihydropyridine-2,6-dicarboxylic acid hydrate, chelidamic acid monohydrate, 4-oxo-1h-pyridine-2,6-dicarboxylic acid hydrate, 1,4-dihydro-4-oxo-2,6-pyridinedicarboxylic acid hydrate, 4-hydroxypyridine-2,6-dicarboxylic acid hydrate, chelidamic acid hydrate, powder, 4-hydroxypyridine-2,6-dicarboxylic acid monohydrate, chelidamic acid hydrate dried material, t , ∼1 mol/mol water
PubChem CID 12302905
IUPAC Name 4-oxo-1H-pyridine-2,6-dicarboxylic acid;hydrate
SMILES C1=C(NC(=CC1=O)C(=O)O)C(=O)O.O

Specifications

Melting Point 263°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 22, 268
Formula Weight 201.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Chelidamic acid monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.