Learn More
Medchemexpress LLC HY-W041489 100mg Medchemexpress, Chelidonic acid CAS:99-32-1 Purity:>98%
SDP
Catalog No. 501934660
Encompass_Preferred
Medchemexpress, HY-W041489 100mg Chelidonic acid CAS:99-32-1 Chelidonic acid is a component of Chelidonium majus L., used as a mild analgesic, an antimicrobial, an acentral nervous system sedative. Chelidonic acid also shows anti-inflammatory activity. Chelidonic acid has potential to inhibit IL-6 production by blocking NF-κB and caspase-1 [1] . Chelidonic acid is a glutamate decarboxylase inhibitor, with a K i of 1.2 μM [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4660 Supplier Medchemexpress LLC Supplier No. HYW041489100MG
Medchemexpress, HY-W041489 100mg Chelidonic acid CAS:99-32-1 Chelidonic acid is a component of Chelidonium majus L., used as a mild analgesic, an antimicrobial, an acentral nervous system sedative. Chelidonic acid also shows anti-inflammatory activity. Chelidonic acid has potential to inhibit IL-6 production by blocking NF-κB and caspase-1 [1] . Chelidonic acid is a glutamate decarboxylase inhibitor, with a K i of 1.2 μM [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.