Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chelidonic acid | 99-32-1 | MFCD00006577 | >98.0% | 184.10 g/mol | C7H4O6 | 1 ML
SDP

Catalog No. 5000208486
Encompass_Preferred

Chelidonic acid is a small-molecule research compound (CAS 99-32-1) reported to modulate NF-κB and caspase-related pathways. The compound is described as a powder with defined solubility and storage recommendations for in vitro and in vivo use.

  • chemical formula: C7H4O6.
  • molecular weight: 184.10 g/mol.
  • purity: >98% (as listed by distributors).
  • solubility: soluble in DMSO (10 mg/mL, with warming/ultrasonic); aqueous solubility approximately 2.5 mg/mL with ultrasonic assistance.
  • recommended in vivo formulation: sequential addition of solvents (10% DMSO → 40% PEG300 → 5% Tween-80 → 45% saline) to obtain a clear solution.
  • storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-08486 Supplier Medchemexpress LLC Supplier No. HYW04148910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chelidonic acid is a small-molecule research compound (CAS 99-32-1) reported to modulate NF-κB and caspase-related pathways. The compound is described as a powder with defined solubility and storage recommendations for in vitro and in vivo use.

  • chemical formula: C7H4O6.
  • molecular weight: 184.10 g/mol.
  • purity: >98% (as listed by distributors).
  • solubility: soluble in DMSO (10 mg/mL, with warming/ultrasonic); aqueous solubility approximately 2.5 mg/mL with ultrasonic assistance.
  • recommended in vivo formulation: sequential addition of solvents (10% DMSO → 40% PEG300 → 5% Tween-80 → 45% saline) to obtain a clear solution.
  • storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.