Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chelidonic Acid Monohydrate 95.0+%, TCI America™
SDP

Catalog No. C09555G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09555G 5 g
C095525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C09555G Supplier TCI America Supplier No. C09555G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6003-94-7
Molecular Formula C7H6O7
Molecular Weight (g/mol) 202.118
MDL Number MFCD00149433
InChI Key KJZJAKSEIQOKAK-UHFFFAOYSA-N
Synonym chelidonic acid monohydrate, compound xi* hydrate, 4-oxo-4h-pyran-2,6-dicarboxylic acid monohydrate, chelidonic acidmonohydrate, chelidonic acid 1-hydrate, c7h4o6.h2o, 2,6-dicarboxy-4-oxo-4h-pyran monohydrate, 4-oxo-4h-pyran-2,6-dicarboxylic acid hydrate, 4h-pyran-2,6-dicarboxylicacid, 4-oxo-, hydrate 1:1
PubChem CID 51346158
IUPAC Name 4-oxopyran-2,6-dicarboxylic acid;hydrate
SMILES C1=C(OC(=CC1=O)C(=O)O)C(=O)O.O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 184.10
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Chelidonic Acid Monohydrate
EINECSNumber (9)-1292
RTECSNumber UP7350000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.