Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S High Performance Precision Pump Tubing, Chem-Durance Bio, L/S 36; 50 ft
Catalog No. NC0554512
Encompass
Applications: Laboratory, pharmaceutical processing, and biotech Characteristics: Excellent chemical resistance, excellent life and durability under pressure, low spallation, plasticizer-free inner liner with elastomer jacket, opaque
Catalog No. NC0554512 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0644236/DEL
Applications: Laboratory, pharmaceutical processing, and biotech Characteristics: Excellent chemical resistance, excellent life and durability under pressure, low spallation, plasticizer-free inner liner with elastomer jacket, opaque