Learn More
eMolecules​ 756526-01-9 | O-(N-Fmoc-2-aminoethyl)-O'-(2-carboxyethyl)undecaethyleneglycol | Chem-Impex | MFCD08064302 | 839.973 | C42H65NO16 | 97.000 | OC(=O)CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(=O)OCC1c2ccccc2-c2ccccc12 | 250mg | 112749397
Catalog No. 5000129039 Shop All eMolecules Inc. Products
Encompass_Preferred
O-(N-Fmoc-2-aminoethyl)-O'-(2-carboxyethyl)undecaethyleneglycol | Chem-Impex | 756526-01-9 | MFCD08064302 | 839.973 | C42H65NO16 | 97.000 | OC(=O)CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(=O)OCC1c2ccccc2-c2ccccc12 | 250mg | 112749397
Catalog No. 50-001-29039 Supplier eMolecules​ Supplier No. 112749397
O-(N-Fmoc-2-aminoethyl)-O'-(2-carboxyethyl)undecaethyleneglycol | Chem-Impex | 756526-01-9 | MFCD08064302 | 839.973 | C42H65NO16 | 97.000 | OC(=O)CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(=O)OCC1c2ccccc2-c2ccccc12 | 250mg | 112749397