Learn More
Pickering Laboratories Inc CHEM LITHIUM DILUENT 4X250ML/C
SDP
Catalog No. NC9726800
Encompass_Preferred
Lithium Diluent, pH 2.36, case of 4 (250 mL/bottle) Amino Acid Analysis.
Catalog No. NC9726800 Supplier Pickering Laboratories Inc Supplier No. LI220
Lithium Diluent, pH 2.36, case of 4 (250 mL/bottle) Amino Acid Analysis.