Learn More
Pickering Laboratories Inc CHEM LITHIUM ELUENT 4X1QT/CS
SDP
Catalog No. NC9730453
Encompass_Preferred
Lithium Eluant, pH 7.50, case of 4 (950 mL/bottle) Amino Acid Analysis.
Catalog No. NC9730453 Supplier Pickering Laboratories Inc Supplier No. LI750
Lithium Eluant, pH 7.50, case of 4 (950 mL/bottle) Amino Acid Analysis.