Learn More
Chemglass Life Sciences Cap, Chem Snap, 11mm, Green, 10mil PTFE Septum
SDP
Catalog No. 50230989
Encompass_Preferred
Cap, Chem Snap, 11mm, Green, 10mil PTFE Septum. Chem Snap Caps™ may be snapped on and off by hand which eliminates the need for metal crimping and decapping tools, thus reducing potential hazards due to sharp aluminum edges. Large opening provides greater target area for needle penetration. Snap caps are color coded for easy sample identification.
Catalog No. 50-230-989 Supplier Chemglass Life Sciences Supplier No. CV3550G011
Cap, Chem Snap, 11mm, Green, 10mil PTFE Septum. Chem Snap Caps™ may be snapped on and off by hand which eliminates the need for metal crimping and decapping tools, thus reducing potential hazards due to sharp aluminum edges. Large opening provides greater target area for needle penetration. Snap caps are color coded for easy sample identification.