Learn More
Chemglass Life Sciences Cap, Chem Snap, 11mm, Green, PTFE/Silicone Septum
SDP
Catalog No. 50231002
Encompass_Preferred
Cap, Chem Snap, 11mm, Green, PTFE/Silicone Septum. Chem Snap Caps™ may be snapped on and off by hand which eliminates the need for metal crimping and decapping tools, thus reducing potential hazards due to sharp aluminum edges. Large opening provides greater target area for needle penetration. Snap caps are color coded for easy sample identification.
Catalog No. 50-231-002 Supplier Chemglass Life Sciences Supplier No. CV3590G011
Cap, Chem Snap, 11mm, Green, PTFE/Silicone Septum. Chem Snap Caps™ may be snapped on and off by hand which eliminates the need for metal crimping and decapping tools, thus reducing potential hazards due to sharp aluminum edges. Large opening provides greater target area for needle penetration. Snap caps are color coded for easy sample identification.