Learn More
Chemglass Life Sciences Chemglass Chem-Thread Glass Fitting, Size 15 (19 mm Tube OD), Accepts 12-15 mm Tubing
SDP
Catalog No. 502733529
Encompass_Preferred
The Chem-Thread CG-350-03 is a complete glass threaded fitting assembly for 19 mm outer-diameter glass tubing. It accepts flexible tubing with 12-15 mm outer diameter and is supplied ready to install for laboratory fluid-transfer applications.
  • Designed for 19 mm tube outer diameter.
  • Accepts 12-15 mm outer-diameter flexible tubing.
  • Externally threaded glass shank for secure connections.
  • Includes a fluoroelastomer o-ring for chemical resistance and sealing.
  • Acetal compression cap provides a tight compression seal.
  • Sold individually as a complete assembly.
Catalog No. 50-273-3529 Supplier Chemglass Life Sciences Supplier No. CG35003
The Chem-Thread CG-350-03 is a complete glass threaded fitting assembly for 19 mm outer-diameter glass tubing. It accepts flexible tubing with 12-15 mm outer diameter and is supplied ready to install for laboratory fluid-transfer applications.
  • Designed for 19 mm tube outer diameter.
  • Accepts 12-15 mm outer-diameter flexible tubing.
  • Externally threaded glass shank for secure connections.
  • Includes a fluoroelastomer o-ring for chemical resistance and sealing.
  • Acetal compression cap provides a tight compression seal.
  • Sold individually as a complete assembly.