Learn More
Sigma Aldrich Fine Chemicals Biosciences Chemat precision spin coat1EA
Catalog No. 502908658
Encompass
These compatible easy-to-use devices provide a convenient step-by-step method for processing metalorganic polymer solutions. A two-stage spin process allows dispensing at low speeds and homogenizing the coating at high speeds. The chucks for these spin-coaters must be ordered separately.
Catalog No. 50-290-8658 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5515621EA
Add to Cart
Add to Cart
Request a Quote
These compatible easy-to-use devices provide a convenient step-by-step method for processing metalorganic polymer solutions. A two-stage spin process allows dispensing at low speeds and homogenizing the coating at high speeds. The chucks for these spin-coaters must be ordered separately.