Learn More
Chembio DPP™ HIV 1/2 Assay
GSA_VASDP
Catalog No. 23999100
Change view
Click to view available options
Type:
HIV 1/2 Assay
Catalog No. Type
23-999-100 HIV 1/2 Assay
1 options
This item is not returnable. View return policy
Catalog No. 23-999-100 Supplier Chembio Supplier No. 6595000
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Patented DPP™ technology for the rapid detection of HIV 1 and HIV 2 antibodies in oral fluid and all blood matrices
  • Patented DPP™ technology for the rapid detection of HIV 1 and HIV 2 antibodies in oral fluid and all blood matrices
  • Intended for use as a point-of-care test to aid in the diagnosis of infection with HIV 1 and HIV 2
  • Sharp and distinctive visual results
  • Excellent sensitivity and specificity
  • Built-in procedural control to human IgG
  • SampleTainer™ bottle: a safe, closed system for handling potentially infectious blood and oral fluid samples
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Type HIV 1/2 Assay
Includes 20 Individually Pouched DPP HIV 1/2 Test Devices with Desiccant pouch; Quick reference instructions (for CLIA Waived); 20 copies of subject information notice; 20 oral fluid swabs; 20 disposable 10μL sample loops; 20 DPP HIV SampleTainer bottle - black cap; DPP HIV running buffer - Green Cap; Product insert for the DPP HIV 1/2 assay