Learn More
Chembio HIV Rapid Test Control Pack
GSA_VASDP
Catalog No. 23999103
Change view
Click to view available options
Quantity:
50 Tests
Catalog No. Quantity
23-999-103 50 Tests
1 options
This item is not returnable. View return policy
Catalog No. 23-999-103 Supplier Chembio Supplier No. 6095490
Add to Cart
Add to Cart
This item is not returnable. View return policy
To be used in conjunction with HIV 1/2 STAT-PAK™ (part number 60-9505-1) and SURE CHECK™ HIV 1/2 (part number 60-9507-0) Assays
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Type HIV Rapid Test Control Pack
Includes 1 HIV-1 Reactive Control 0.25 mL,1 HIV-2 Reactive Control 0.25 mL,1 HIV Nonreactive Control 0.25 mL
Quantity 50 Tests