Learn More
Chemglass Life Sciences CHEMcell Rocker BioReactor, Base ONLY
SDP
Supplier:  Chemglass Life Sciences CLS1200100
Encompass
Base Unit Features.Adjustable tray angle.Variable speed rocking.Quiet operation.Optional vent heater.Two-year warranty.Tray Features.22 and 50 liter tray sizes.Stainless steel construction.Easy bag installation.Includes heater and thermocouple. CHEMcell Modular Benchtop Bioreactor System consists of a heavy-duty base unit and a choice of stainless steel rocker trays.Engineered for durability, it can be used in both research and development as well as production environments.The smooth rocking motion provides gentle, efficient mixing and gas transfer.Designed for optimal performance with Chemglass 8-port and 6-port cell culture bags but easily accepts other manufacturers’ bags having the same dimensions.The optional bag and vent heater, made for use with both size trays, keeps vent filters free of harmful condensation that can clog filters and compromise cultures.Click the image below for Rocker BioReactor Systems:
Catalog No. 50-194-1153
Only null left
Add to Cart
Request a Quote