Learn More
Justrite™ ChemCor™ SpillSlope™ Steel Shelf for 23 Gallon Under Fume Hood Safety Cabinet
Catalog No. 19822480 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
23 gal. Under Fumehood Cabinet
Catalog No. For Use With (Equipment)
19-822-480 23 gal. Under Fumehood Cabinet
1 options
Catalog No. 19-822-480 Supplier Justrite™ Supplier No. 29940
  • $738.00
    $501.65 / Each of 1
    Save $236.35
    32% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Increase the efficiency and space of existing cabinetry. The Justrite™ ChemCor™ SpillSlope™ Steel Shelf for 23 Gallon Under Fume Hood Safety Cabinet is the perfect product to add additional storage space. The Chemchor™ thermoplastic liner offers superior protection.
  • Shelves have the ChemCor™ thermoplastic coating and provide superior protection against corrosion from drips and off-gassing from harsh chemicals
  • Patented SpillSlope™ galvanized steel shelves direct spills to back and bottom of a leakproof sump.
Specifications
For Use With (Equipment) 23 gal. Under Fumehood Cabinet
Material Steel
Width (English) Exterior 32.5 in.
Depth (English) Exterior 18 in.