Learn More
Amsco Scientific Steris Verify Valueline Steam Chemical Indicators, 250 strips/pk
Catalog No. NC1869529
Encompass_Preferred
The VERIFY Valueline STEAM Chemical Indicators are Type-1 process indicators as defined by ANSI/AAMI/ISO 11140-1, which through a visible color change from white to black, indicate exposer to a steam sterilization process in an autoclave.
This item is not returnable. View return policy
Catalog No. NC1869529 Supplier Amsco Scientific Steris Supplier No. PCC009
The VERIFY Valueline STEAM Chemical Indicators are Type-1 process indicators as defined by ANSI/AAMI/ISO 11140-1, which through a visible color change from white to black, indicate exposer to a steam sterilization process in an autoclave.